Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1304810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107314.1 | NP_001100784.1 | 6 | 1429 | ||
XM_006253222.1 | XP_006253284.1 | 6 | 1261 | |||
XM_006253223.2 | XP_006253285.1 | 6 | 3003 | |||
XM_008771300.1 | XP_008769522.1 | 5 | 1360 | |||
XM_008771301.1 | XP_008769523.1 | 2 | 1389 |