Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107342.1 | NP_001100812.1 | 7, 8 | 953 | ||
XM_006253729.2 | XP_006253791.1 | 8, 9 | 2975 |