Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004083.1 | NP_001004083.1 | 2 | 565 | ||
XM_006228574.2 | XP_006228636.1 | 1 | 410 | |||
XM_006228575.2 | XP_006228637.1 | 2 | 614 | |||
GenBank mRNA | AY229997.1 | - | 1 | 295 | ||
AY952204.1 | - | 1 | 295 | |||
BC078907.1 | - | 2 | 565 | 7113703 | ||
BC081858.1 | - | 2 | 588 | 7107517 |