Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760022.1 | XP_008758244.1 | 4 | 559 | ||
XM_008760023.1 | XP_008758245.1 | 3 | 511 | |||
XM_008760024.1 | XP_008758246.1 | 4 | 1841 | |||
XM_008760025.1 | XP_008758247.1 | 4 | 543 | |||
XM_219476.7 | XP_219476.5 | 3 | 2044 |