Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107603.1 | NP_001101073.1 | 4 | 1240 | ||
GenBank mRNA | FQ235302.1 | - | 4 | 1232 |