Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004720.2 | NP_001004720.1 | 2 | 371 | ||
NM_001004722.3 | NP_001004722.1 | 3 | 481 | |||
NM_003581.4 | NP_003572.2 | 3 | 481 | |||
XM_006712797.3 | XP_006712860.1 | 3 | 1137 | |||
XM_011511991.2 | XP_011510293.1 | 3 | 575 | |||
XM_011511992.2 | XP_011510294.2 | 2 | 425 | |||
XM_017005103.1 | XP_016860592.1 | 4 | 739 | |||
XM_017005104.1 | XP_016860593.1 | 3 | 751 | |||
XM_017005105.1 | XP_016860594.1 | 3 | 488 | |||
GenBank mRNA | AF043119.1 | - | 1 | 233 | ||
AF047487.1 | - | 2 | 337 | |||
BC000103.2 | - | 2 | 255 | 3505427 | ||
BC007195.1 | - | 2 | 262 | 3608149 | ||
BG480643.1 | - | 3 | 483 | |||
CB959523.1 | - | 300 | ||||
DB456958.1 | - | 2 | 368 | |||
KU178418.1 | - | 1 | 205 | |||
KU178419.1 | - | 1 | 205 |