Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191567.1 | NP_001178496.1 | 13 | 1889 | ||
XM_006232499.2 | XP_006232561.1 | 12 | 2359 | |||
XM_006232500.1 | XP_006232562.1 | 11 | 2272 | |||
GenBank mRNA | BC091440.1 | - | 5 | 843 |