Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16142
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
12
1695
NM_001220474.2
NP_001207403.1
11
1641
NM_001220475.2
NP_001207404.1
12
1507
NM_001220476.2
NP_001207405.1
12
1507
NM_001220477.2
NP_001207406.1
10
1514
NM_001220478.2
NP_001207407.1
9
1439
NM_001220479.2
NP_001207408.1
3
256
NM_001220480.2
NP_001207409.1
3
256
NM_001282536.1
NP_001269465.1
10
1202
NM_003625.4
NP_003616.2
13
1558
XM_006719670.3
XP_006719733.1
10
8750
XM_011538906.2
XP_011537208.1
11
1635
XM_011538907.2
XP_011537209.1
11
1635
XM_017020079.1
XP_016875568.1
13
1542
XM_017020080.1
XP_016875569.1
12
1689
XM_017020081.1
XP_016875570.1
12
1689
XM_017020082.1
XP_016875571.1
13
1542
XM_017020083.1
XP_016875572.1
12
1689
XM_017020084.1
XP_016875573.1
12
1689
XM_017020085.1
XP_016875574.1
12
1689
XM_017020086.1
XP_016875575.1
13
1542
XM_017020087.1
XP_016875576.1
12
1676
XM_017020088.1
XP_016875577.1
12
1922
XM_017020089.1
XP_016875578.1
11
1635
XM_017020090.1
XP_016875579.1
13
1542
XM_017020091.1
XP_016875580.1
12
1689
XM_017020092.1
XP_016875581.1
12
1689
XM_017020093.1
XP_016875582.1
12
1689
XM_017020094.1
XP_016875583.1
12
1467
XM_017020095.1
XP_016875584.1
12
1689
XM_017020096.1
XP_016875585.1
11
1614
XM_017020097.1
XP_016875586.1
13
1542
XM_017020098.1
XP_016875587.1
13
1542
XM_017020099.1
XP_016875588.1
11
1635
XM_017020100.1
XP_016875589.1
12
1689
XM_017020101.1
XP_016875590.1
13
1542
XM_017020102.1
XP_016875591.1
11
1601
XM_017020103.1
XP_016875592.1
12
1689
XM_017020104.1
XP_016875593.1
12
1689
XM_017020105.1
XP_016875594.1
11
1614
XM_017020106.1
XP_016875595.1
12
1689
XM_017020107.1
XP_016875596.1
13
1542
XM_017020108.1
XP_016875597.1
13
1542
XM_017020109.1
XP_016875598.1
12
1689
XM_017020110.1
XP_016875599.1
11
1847
XM_017020111.1
XP_016875600.1
12
1689
XM_017020112.1
XP_016875601.1
10
1514
XM_017020113.1
XP_016875602.1
10
1514
XM_017020114.1
XP_016875603.1
10
1514
XM_017020115.1
XP_016875604.1
10
1514
XM_017020116.1
XP_016875605.1
10
1514
XM_017020117.1
XP_016875606.1
9
1249
XM_017020118.1
XP_016875607.1
11
1489
XM_017020119.1
XP_016875608.1
10
1514
XM_017020120.1
XP_016875609.1
10
1202
GenBank mRNA
AB210009.1
-
12
1693
AF034799.1
-
13
1566
AK123372.1
-
3
191
AK126971.1
-
11
1601
AK294505.1
-
9
1249
AK296380.1
-
3
227
AK299853.1
-
10
1463
AK302658.1
-
10
1173
BC104912.1
-
12
1491
8143915
BC143485.1
-
12
1491
9051993
DA533482.1
-
4
313
DA687865.1
-
3
240
DC322478.1
-
3
256
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0