Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PKP4 SIRNA Assays ›
Gene Symbol:
PKP4
Gene Name:
plakophilin 4
Gene Aliases:
p0071
Chromosome Location:
Chr.2: 158456880 - 158681429 on Build GRCh38
UniGene ID:
Hs.407580
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16149
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PKP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005476.2
NP_001005476.1
2
456
NM_001304969.1
NP_001291898.1
2
456
NM_001304970.1
NP_001291899.1
2
456
NM_001304971.1
NP_001291900.1
2
456
NM_003628.4
NP_003619.2
2
456
XM_011512016.1
XP_011510318.1
2
893
XM_011512017.1
XP_011510319.1
2
203
XM_011512018.1
XP_011510320.1
2
893
XM_011512019.1
XP_011510321.1
2
203
XM_011512020.2
XP_011510322.1
2
240
XM_011512021.2
XP_011510323.1
2
240
XM_011512022.1
XP_011510324.1
2
521
XM_011512026.2
XP_011510328.1
2
240
XM_017005122.1
XP_016860611.1
2
893
XM_017005123.1
XP_016860612.1
2
1428
XM_017005124.1
XP_016860613.1
2
893
XM_017005125.1
XP_016860614.1
2
240
XM_017005126.1
XP_016860615.1
2
203
XM_017005127.1
XP_016860616.1
2
893
XM_017005128.1
XP_016860617.1
2
240
XM_017005129.1
XP_016860618.1
2
521
XM_017005130.1
XP_016860619.1
2
521
XM_017005133.1
XP_016860622.1
2
410
XM_017005134.1
XP_016860623.1
2
893
GenBank mRNA
AK054911.1
-
2
970
AK055823.1
-
548
BC048013.1
-
2
372
X81889.1
-
2
253
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell junction assembly
cell-cell signaling
single organismal cell-cell adhesion
regulation of cell adhesion
positive regulation of cytokinesis
positive regulation of GTPase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0