Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175765.2 | NP_786941.1 | 3 | 347 | ||
XM_006238353.2 | XP_006238415.1 | 4 | 571 | |||
XM_006238354.2 | XP_006238416.1 | 4 | 570 | |||
XM_008763779.1 | XP_008762001.1 | 4 | 570 | |||
GenBank mRNA | AB285525.1 | - | 250 | |||
AF339084.1 | - | 3 | 347 | |||
AF372090.1 | - | 3 | 347 | |||
BC093606.1 | - | 4 | 505 | 7317952 |