Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764393.1 | XP_008762615.1 | 21 | 3756 | ||
XM_008764394.1 | XP_008762616.1 | 21 | 2813 | |||
XM_008764395.1 | XP_008762617.1 | 21 | 2651 | |||
XM_008764396.1 | XP_008762618.1 | 21 | 2804 | |||
XM_008764397.1 | XP_008762619.1 | 22 | 2827 | |||
XM_008764398.1 | XP_008762620.1 | 21 | 3755 |