Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cptp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007703.1 | NP_001007704.1 | 3 | 464 | ||
XM_006239576.2 | XP_006239638.1 | 3 | 625 | |||
GenBank mRNA | BC083599.1 | - | 3 | 464 | 7133008 |