Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014078.1 | NP_001014100.1 | 2 | 365 | ||
XM_006240232.2 | XP_006240294.1 | 3 | 664 | |||
XM_006240233.2 | XP_006240295.1 | 3 | 404 | |||
XM_006240235.2 | XP_006240297.2 | 2 | 361 | |||
GenBank mRNA | BC089849.1 | - | 2 | 365 | 7375058 |