Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kdelc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025123.3 | NP_001020294.3 | 2 | 257 | ||
GenBank mRNA | BC093594.1 | - | 2 | 257 | 7314531 | |
BC100617.1 | - | 2 | 256 | 7934552 | ||
FQ211817.1 | - | 2 | 158 |