Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304472.1 | NP_001291401.1 | 3 | 550 | ||
NM_001304473.1 | NP_001291402.1 | 4 | 605 | |||
NM_001304474.1 | NP_001291403.1 | 3 | 550 | |||
NM_003741.3 | NP_003732.2 | 3 | 550 | |||
NR_130747.1 | 3 | 550 | ||||
XM_011513254.1 | XP_011511556.1 | 3 | 403 | |||
XM_011513256.1 | XP_011511558.1 | 2 | 191 | |||
XM_017007388.1 | XP_016862877.1 | 3 | 403 | |||
XM_017007389.1 | XP_016862878.1 | 3 | 403 | |||
XM_017007390.1 | XP_016862879.1 | 3 | 403 | |||
XM_017007391.1 | XP_016862880.1 | 3 | 403 | |||
XM_017007392.1 | XP_016862881.1 | 3 | 403 | |||
XM_017007393.1 | XP_016862882.1 | 3 | 403 | |||
XM_017007394.1 | XP_016862883.1 | 2 | 191 | |||
GenBank mRNA | AB208947.1 | - | 3 | 323 | ||
AF209928.1 | - | 3 | 550 | |||
AF209929.1 | - | 4 | 601 | |||
AF209930.1 | - | 4 | 605 | |||
AF283325.1 | - | 3 | 550 | |||
AK074147.1 | - | 3 | 303 | |||
AK293456.1 | - | 3 | 272 | |||
AK295632.1 | - | 3 | 190 | |||
AK296316.1 | - | 2 | 149 | |||
AY358926.1 | - | 3 | 440 | |||
BC112345.1 | - | 3 | 335 | 40008417 |