Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199111.1 | NP_954542.1 | 2, 3 | 654 | ||
GenBank mRNA | BC061795.1 | - | 2, 3 | 654 | 5597840 |