Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025712.1 | NP_001020883.1 | 5 | 673 | ||
GenBank mRNA | BC099174.1 | - | 5 | 673 | 7382658 | |
FQ216632.1 | - | 5 | 674 | |||
FQ217144.1 | - | 5 | 692 | |||
FQ224887.1 | - | 5 | 692 |