Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108247.2 | NP_001101717.1 | 1 | 232 | ||
XM_006256719.2 | XP_006256781.1 | 2 | 773 | |||
XM_006256720.2 | XP_006256782.1 | 2 | 1030 | |||
XM_008773075.1 | XP_008771297.1 | 2 | 1001 | |||
GenBank mRNA | AF452730.1 | - | 2 | 980 | ||
BC167040.1 | - | 1 | 232 | 7135266 |