Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108250.1 | NP_001101720.1 | 4, 5 | 558 | ||
XM_006256731.2 | XP_006256793.1 | 3, 4 | 621 | |||
GenBank mRNA | AB232988.1 | - | 4, 5 | 582 | ||
AB232989.1 | - | 3, 4 | 500 | |||
AB232990.1 | - | 4, 5 | 582 |