Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271190.1 | NP_001258119.1 | 21 | 2877 | ||
XM_006257525.2 | XP_006257587.2 | 22 | 2879 | |||
XM_006257526.2 | XP_006257588.2 | 22 | 2894 | |||
XM_006257527.2 | XP_006257589.2 | 22 | 2894 | |||
XM_008773524.1 | XP_008771746.1 | 22 | 2894 | |||
XM_008773526.1 | XP_008771748.1 | 22 | 2847 |