Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001515.1 | NP_001001515.1 | 4, 5 | 311 | ||
XM_006252397.2 | XP_006252459.1 | 4, 5 | 547 | |||
XM_006252399.2 | XP_006252461.1 | 4, 5 | 543 | |||
XM_006252400.2 | XP_006252462.1 | 4, 5 | 543 | |||
XM_006252401.2 | XP_006252463.1 | 4, 5 | 543 | |||
XM_008770874.1 | XP_008769096.1 | 5, 6 | 443 | |||
XM_008770875.1 | XP_008769097.1 | 4, 5 | 548 | |||
XM_008770876.1 | XP_008769098.1 | 4, 5 | 548 | |||
XM_008770877.1 | XP_008769099.1 | 4, 5 | 547 | |||
XM_008770878.1 | XP_008769100.1 | 2, 3 | 1009 | |||
XM_008770879.1 | XP_008769101.1 | 4, 5 | 547 | |||
XM_008770880.1 | XP_008769102.1 | 2, 3 | 236 | |||
GenBank mRNA | AY533473.1 | - | 4, 5 | 311 | ||
AY533474.1 | - | 4, 5 | 311 | |||
AY609384.1 | - | 1, 2 | 326 |