Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003827.3 | NP_003818.2 | 5 | 701 | ||
NR_038456.1 | 5 | 701 | ||||
NR_038457.1 | 3 | 504 | ||||
XM_011527436.1 | XP_011525738.1 | 5 | 375 | |||
XM_011527437.1 | XP_011525739.1 | 4 | 454 | |||
GenBank mRNA | AK098401.1 | - | 5 | 534 | ||
AK126519.1 | - | 2 | 1588 | |||
AK292244.1 | - | 5 | 465 | |||
AK308633.1 | - | 6 | 591 | |||
AK310114.1 | - | 3 | 268 | |||
BC001165.1 | - | 5 | 473 | 3533668 | ||
BC007432.2 | - | 5 | 464 | 2962847 | ||
BC028234.1 | - | 5 | 519 | 5245770 | ||
BC091511.1 | - | 5 | 480 | 30512911 | ||
BQ948081.1 | - | 5 | 683 | |||
BT019568.1 | - | 5 | 383 | |||
CX873074.1 | - | 3 | 246 | |||
U39412.1 | - | 5 | 450 |