Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108497.1 | NP_001101967.1 | 5 | 807 | ||
XM_006229987.1 | XP_006230049.1 | 6 | 696 | |||
XM_006229988.2 | XP_006230050.1 | 6 | 627 | |||
XM_006229989.1 | XP_006230051.1 | 6 | 666 | |||
XM_006229990.1 | XP_006230052.1 | 6 | 654 | |||
XM_006229991.1 | XP_006230053.1 | 5 | 615 | |||
XM_006229992.1 | XP_006230054.1 | 5 | 540 |