Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004278.3 | NP_001004278.2 | 1 | 138 | ||
NM_001270736.1 | NP_001257665.1 | 1 | 138 | |||
NM_001270737.1 | NP_001257666.1 | 1 | 138 | |||
XM_006230847.2 | XP_006230909.1 | 1 | 413 | |||
XM_008760155.1 | XP_008758377.1 | 1 | 414 | |||
XM_008760156.1 | XP_008758378.1 | 1 | 413 | |||
XM_008760157.1 | XP_008758379.1 | 1 | 415 | |||
GenBank mRNA | AY690667.1 | - | 1 | 155 | ||
BC097254.1 | - | 1 | 119 | 7453704 |