Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108526.1 | NP_001101996.1 | 11 | 1758 | ||
XM_006231528.2 | XP_006231590.1 | 12 | 1870 |