Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101669.1 | NP_001095139.1 | 5 | 616 | ||
NM_003866.2 | NP_003857.2 | 6 | 715 | |||
XM_011532391.2 | XP_011530693.1 | 5 | 700 | |||
XM_011532392.2 | XP_011530694.1 | 5 | 466 | |||
XM_017008794.1 | XP_016864283.1 | 5 | 700 | |||
XM_017008795.1 | XP_016864284.1 | 5 | 700 | |||
XM_017008796.1 | XP_016864285.1 | 5 | 700 | |||
GenBank mRNA | AK302936.1 | - | 4 | 382 | ||
AY753912.1 | - | 2 | 120 | |||
BC005273.1 | - | 4 | 493 | |||
BC005274.1 | - | 5 | 391 | |||
BC072447.1 | - | 4 | 523 | |||
BC110918.1 | - | 4 | 473 | 6160633 | ||
BC133005.1 | - | 2 | 171 | 40147027 | ||
BC144255.1 | - | 2 | 171 | |||
BX649090.1 | - | 6 | 556 | |||
U96922.1 | - | 2 | 241 |