Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139486.1 | NP_001132958.1 | 9 | 853 | ||
XM_006232976.2 | XP_006233038.1 | 7 | 842 | |||
GenBank mRNA | AY724489.1 | - | 9 | 784 | ||
DY546285.1 | - | 5 | 415 |
Set Membership: |
![]() |