Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108817.2 | NP_001102287.2 | 2, 3 | 323 | ||
XM_006257078.1 | XP_006257140.1 | 2, 3 | 392 | |||
XM_006257079.1 | XP_006257141.1 | 2, 3 | 392 | |||
GenBank mRNA | BP500254.1 | - | 2, 3 | 329 |