Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191890.1 | NP_001178819.1 | 4 | 553 | ||
XM_006246767.2 | XP_006246829.1 | 5 | 605 | |||
GenBank mRNA | FQ223142.1 | - | 4 | 944 |