Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163277.1 | NP_001156749.1 | 6 | 1341 | ||
XM_008760004.1 | XP_008758226.1 | 10 | 2436 |