Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003969.3 | NP_003960.1 | 2 | 733 | ||
GenBank mRNA | AB012191.1 | - | 2 | 138 | ||
AF075599.1 | - | 2 | 141 | |||
BC058924.1 | - | 2 | 247 | 6162220 | ||
BT006754.1 | - | 2 | 138 | |||
BU508830.1 | - | 2 | 677 |
Gene Symbol: UBE2MP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002837.1 | 1 | 287 | |||
GenBank mRNA | AW376368.1 | - | 1 | 89 |