Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTPIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321135.1 | NP_001308064.1 | 7, 8 | 1007 | ||
NM_001321136.1 | NP_001308065.1 | 8, 9 | 1226 | |||
NM_001321137.1 | NP_001308066.1 | 8, 9 | 823 | |||
NM_003978.4 | NP_003969.2 | 7, 8 | 1007 | |||
NR_135552.1 | 7, 8 | 1007 | ||||
XM_006720737.2 | XP_006720800.1 | 7, 8 | 739 | |||
XM_011522163.1 | XP_011520465.1 | 8, 9 | 1025 | |||
XM_011522164.1 | XP_011520466.1 | 8, 9 | 693 | |||
XM_011522165.2 | XP_011520467.1 | 7, 8 | 548 | |||
XM_011522166.1 | XP_011520468.1 | 8, 9 | 1025 | |||
XM_011522167.1 | XP_011520469.1 | 8, 9 | 1025 | |||
XM_011522168.2 | XP_011520470.1 | 8, 9 | 1025 | |||
XM_011522169.1 | XP_011520471.1 | 8, 9 | 1025 | |||
GenBank mRNA | AB451310.1 | - | 7, 8 | 518 | ||
AB451440.1 | - | 7, 8 | 518 | |||
AF038602.1 | - | 7, 8 | 518 | |||
AF038603.1 | - | 7, 8 | 518 | |||
AK298733.1 | - | 7, 8 | 760 | |||
AK304052.1 | - | 7, 8 | 796 | |||
AK308462.1 | - | 8, 9 | 823 | |||
AK309856.1 | - | 8, 9 | 1189 | |||
AK310072.1 | - | 8, 9 | 940 | |||
AK316220.1 | - | 7, 8 | 770 | |||
BC008602.1 | - | 7, 8 | 879 | 4180398 | ||
KU178458.1 | - | 7, 8 | 518 | |||
KU178459.1 | - | 7, 8 | 518 | |||
U94778.1 | - | 7, 8 | 734 |