Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-M10-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008851.1 | NP_001008851.1 | 3 | 502 | ||
XM_006255888.2 | XP_006255950.1 | 3 | 543 |