Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009517.1 | NP_001009517.1 | 1 | 613 | ||
GenBank mRNA | AY510342.1 | - | 1 | 613 |
Gene Symbol: Vom1r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009516.1 | NP_001009516.1 | 1 | 613 | ||
GenBank mRNA | AY510338.1 | - | 1 | 613 |