Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134614.1 | NP_001128086.1 | 3, 4 | 505 | ||
XM_006232145.2 | XP_006232207.1 | 3, 4 | 1109 | |||
XM_006232146.2 | XP_006232208.1 | 3, 4 | 638 | |||
XM_006232147.2 | XP_006232209.1 | 2, 3 | 306 | |||
XM_006232148.2 | XP_006232210.1 | 2, 3 | 306 | |||
GenBank mRNA | EU733648.1 | - | 3, 4 | 406 |