siRNA Details
Gene Symbol: Hoxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257706.2 | XP_006257768.1 | 5 | 6866 | ||
XM_006257707.2 | XP_006257769.1 | 3 | 1163 | |||
XM_008773712.1 | XP_008771934.1 | 4 | 4896 |
Gene Symbol: NEWGENE_1561431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236526.2 | XP_006236588.1 | 6866 | |||
XM_006236527.2 | XP_006236589.1 | 3 | 1163 | |||
XM_008762939.1 | XP_008761161.1 | 4 | 4896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
