Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280558.1 | NP_001267487.1 | 6 | 798 | ||
NM_006032.3 | NP_006023.1 | 5 | 786 | |||
XM_005268216.1 | XP_005268273.1 | 6 | 742 | |||
XM_005268217.1 | XP_005268274.1 | 4 | 436 | |||
GenBank mRNA | AB009288.1 | - | 5 | 586 | ||
AK125195.1 | - | 4 | 1348 | |||
AK222675.1 | - | 5 | 546 | |||
AK293655.1 | - | 6 | 798 | |||
AK294460.1 | - | 4 | 701 | |||
AK314182.1 | - | 5 | 599 | |||
AL137688.1 | - | 3 | 1997 | |||
BC018046.1 | - | 5 | 592 | 4799650 |