Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN2 SIRNA Assays ›
Gene Symbol:
NRXN2
Gene Name:
neurexin 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 64606174 - 64723188 on Build GRCh38
UniGene ID:
Hs.372938
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17949
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015080.3
NP_055895.1
23
4728
NM_138732.2
NP_620060.1
20
4518
NM_138734.2
NP_620063.1
7
1640
XM_005274400.3
XP_005274457.1
22
4260
XM_011545370.1
XP_011543672.1
22
4290
XM_011545371.1
XP_011543673.1
22
4287
XM_011545373.1
XP_011543675.1
22
4269
XM_011545375.1
XP_011543677.1
21
4245
XM_011545385.1
XP_011543687.1
15
2635
XM_017018563.1
XP_016874052.1
21
4260
XM_017018564.1
XP_016874053.1
21
4257
XM_017018565.1
XP_016874054.1
21
4254
XM_017018566.1
XP_016874055.1
21
4218
XM_017018567.1
XP_016874056.1
21
4200
XM_017018568.1
XP_016874057.1
21
4191
XM_017018569.1
XP_016874058.1
20
4188
XM_017018572.1
XP_016874061.1
22
3585
XM_017018574.1
XP_016874063.1
21
3555
XM_017018575.1
XP_016874064.1
21
3552
XM_017018576.1
XP_016874065.1
17
3200
XM_017018577.1
XP_016874066.1
15
2626
GenBank mRNA
AB023138.2
-
21
4495
AB035266.1
-
22
4380
AB177862.1
-
19
3432
AK098211.1
-
3
1337
BC150275.1
-
21
4495
9007249
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053846, XM_008760061, XM_008760064, XM_008760058, XM_008760060, and others.This siRNA also targets the following transcripts in Mouse: NM_001205234, AK147665, BM944179, BC060272, AK147295.
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
signal transduction
chemical synaptic transmission
neurotransmitter secretion
synapse assembly
adult behavior
social behavior
vocal learning
vocalization behavior
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
transmembrane signaling receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0