Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109296.1 | NP_001102766.1 | 7 | 816 | ||
XM_008766561.1 | XP_008764783.1 | 8 | 1276 | |||
XM_008766562.1 | XP_008764784.1 | 6 | 693 | |||
GenBank mRNA | FQ233502.1 | - | 8 | 880 |