Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raet1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758633.1 | XP_008756855.1 | 3 | 830 |
Gene Symbol: Rrlt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161691.1 | NP_001155163.1 | 752 | |||
GenBank mRNA | FJ971664.1 | - | 752 |