Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004271.3 | NP_004262.1 | 1, 2 | 168 | ||
XM_017011505.1 | XP_016866994.1 | 1, 2 | 230 | |||
GenBank mRNA | AF057178.2 | - | 1, 2 | 157 | ||
AY931019.1 | - | 1, 2 | 134 | |||
BC038846.1 | - | 1, 2 | 178 | 6015123 | ||
CD695164.1 | - | 1, 2 | 232 |