Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760965.1 | XP_008759187.1 | 4, 5 | 228 |
Gene Symbol: LOC678817 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760981.1 | XP_008759203.1 | 538 |