Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC678893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750190.3 | XP_003750238.1 | 5 | 1237 | ||
XM_008764871.1 | XP_008763093.1 | 3 | 3164 | |||
XM_008764872.1 | XP_008763094.1 | 5 | 3434 | |||
XM_008764873.1 | XP_008763095.1 | 6 | 3959 | |||
XM_008764874.1 | XP_008763096.1 | 6 | 1624 | |||
XM_008764875.1 | XP_008763097.1 | 5 | 1500 | |||
XM_008764876.1 | XP_008763098.1 | 5 | 1746 | |||
XM_008764877.1 | XP_008763099.1 | 6 | 3497 |