Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080395.2 | NP_001073864.2 | 7 | 814 | ||
NM_004920.2 | NP_004911.2 | 6 | 592 | |||
XM_006722193.2 | XP_006722256.1 | 7 | 807 | |||
XM_006722194.2 | XP_006722257.1 | 7 | 809 | |||
XM_006722195.2 | XP_006722258.1 | 5 | 601 | |||
XM_006722196.2 | XP_006722259.1 | 7 | 776 | |||
XM_011525509.2 | XP_011523811.1 | 6 | 2243 | |||
XM_011525510.2 | XP_011523812.1 | 6 | 619 | |||
XM_011525511.2 | XP_011523813.1 | 2 | 472 | |||
XM_017025421.1 | XP_016880910.1 | 6 | 606 | |||
XM_017025422.1 | XP_016880911.1 | 6 | 765 | |||
GenBank mRNA | AB014541.2 | - | 6 | 592 | ||
AK131395.1 | - | 5 | 605 | |||
AK131529.1 | - | 7 | 814 |