Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS6 SIRNA Assays ›
Gene Symbol:
RGS6
Gene Name:
regulator of G-protein signaling 6
Gene Aliases:
GAP
Chromosome Location:
Chr.14: 71932100 - 72632478 on Build GRCh38
UniGene ID:
Hs.509872
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18504
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204416.2
NP_001191345.1
8
783
NM_001204417.2
NP_001191346.1
8
783
NM_001204418.2
NP_001191347.1
8
783
NM_001204419.2
NP_001191348.1
8
783
NM_001204420.2
NP_001191349.1
8
783
NM_001204421.2
NP_001191350.1
8
783
NM_001204422.2
NP_001191351.1
8
783
NM_001204423.1
NP_001191352.1
7
548
NM_001204424.1
NP_001191353.1
8
1021
NM_004296.6
NP_004287.3
8
783
NR_135235.1
8
783
XM_011537393.2
XP_011535695.1
8
1168
XM_011537397.1
XP_011535699.1
6
338
XM_011537407.2
XP_011535709.1
8
1059
XM_017021818.1
XP_016877307.1
8
683
XM_017021819.1
XP_016877308.1
8
683
XM_017021820.1
XP_016877309.1
7
518
XM_017021821.1
XP_016877310.1
7
518
XM_017021822.1
XP_016877311.1
7
518
XM_017021823.1
XP_016877312.1
7
518
XM_017021824.1
XP_016877313.1
8
683
XM_017021825.1
XP_016877314.1
7
518
XM_017021826.1
XP_016877315.1
7
518
XM_017021827.1
XP_016877316.1
7
518
XM_017021828.1
XP_016877317.1
7
518
XM_017021829.1
XP_016877318.1
8
683
XM_017021830.1
XP_016877319.1
7
518
XM_017021831.1
XP_016877320.1
7
518
XM_017021832.1
XP_016877321.1
7
518
XM_017021833.1
XP_016877322.1
7
518
XM_017021834.1
XP_016877323.1
6
338
GenBank mRNA
AB209462.1
-
8
589
AF073921.1
-
3
146
AF107619.1
-
7
498
AF107620.1
-
7
498
AF156932.1
-
7
498
AK294486.1
-
8
1021
AK294895.1
-
7
548
AK302319.1
-
6
394
AK310463.1
-
8
681
AK313481.1
-
8
843
AY309097.1
-
7
498
AY309098.1
-
7
498
AY309099.1
-
7
498
AY309100.1
-
7
498
BC104877.1
-
8
676
8143880
BC113572.1
-
8
676
8322624
DA386861.1
-
6
357
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0