Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304447.1 | NP_001291376.1 | 7 | 1785 | ||
NM_014653.3 | NP_055468.2 | 6 | 1954 | |||
XM_017020243.1 | XP_016875732.1 | 7 | 2239 | |||
XM_017020244.1 | XP_016875733.1 | 8 | 2400 | |||
XM_017020245.1 | XP_016875734.1 | 8 | 1897 | |||
XM_017020246.1 | XP_016875735.1 | 6 | 1628 | |||
XM_017020247.1 | XP_016875736.1 | 7 | 1771 | |||
XM_017020248.1 | XP_016875737.1 | 7 | 2243 | |||
XM_017020249.1 | XP_016875738.1 | 6 | 2100 | |||
XM_017020250.1 | XP_016875739.1 | 4 | 1654 | |||
XM_017020251.1 | XP_016875740.1 | 5 | 1465 | |||
XM_017020252.1 | XP_016875741.1 | 5 | 1943 | |||
GenBank mRNA | AB018332.2 | - | 6 | 1691 | ||
AK293764.1 | - | 5 | 1213 | |||
BC037969.1 | - | 1 | 180 | |||
BC110330.1 | - | 6 | 1737 | 5735001 | ||
BC136687.1 | - | 5 | 1483 | 9020677 | ||
BC136688.1 | - | 5 | 1483 | 9020678 | ||
BX647283.1 | - | 7 | 1736 |