Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001068704.5 | XP_001068704.3 | 6 | 1039 | ||
XM_006249947.2 | XP_006250009.1 | 6 | 965 | |||
XM_006249948.2 | XP_006250010.1 | 6 | 1040 | |||
XM_006249949.2 | XP_006250011.1 | 5 | 862 | |||
XM_006249950.2 | XP_006250012.1 | 4 | 679 | |||
XM_006249951.2 | XP_006250013.1 | 6 | 1040 | |||
GenBank mRNA | FQ211198.1 | - | 5 | 842 |