Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18879
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
15
1856
NM_001205119.1
NP_001192048.1
15
1899
NM_001205120.1
NP_001192049.1
14
1800
NM_001205121.1
NP_001192050.1
14
1740
NM_001205122.1
NP_001192051.1
12
1607
NM_014741.4
NP_055556.2
14
1745
XM_005253262.2
XP_005253319.1
16
1695
XM_005253263.2
XP_005253320.1
16
1986
XM_005253265.2
XP_005253322.1
17
2192
XM_005253266.2
XP_005253323.1
16
1921
XM_005253268.2
XP_005253325.1
14
1298
XM_006718394.2
XP_006718457.1
17
2042
XM_006718395.2
XP_006718458.1
17
1977
XM_006718396.2
XP_006718459.1
16
2076
XM_011520493.1
XP_011518795.1
18
2179
XM_011520496.2
XP_011518798.1
15
1954
XM_011520499.2
XP_011518801.1
15
1568
XM_011520500.1
XP_011518802.1
17
2015
XM_011520501.1
XP_011518803.1
16
1943
XM_011520502.1
XP_011518804.1
16
1916
XM_011520503.1
XP_011518805.1
15
1832
XM_017018599.1
XP_016874088.1
17
2127
XM_017018600.1
XP_016874089.1
16
1668
XM_017018601.1
XP_016874090.1
15
1843
XM_017018602.1
XP_016874091.1
16
2093
XM_017018603.1
XP_016874092.1
17
2210
XM_017018604.1
XP_016874093.1
15
1887
XM_017018605.1
XP_016874094.1
16
1878
XM_017018606.1
XP_016874095.1
15
1822
XM_017018607.1
XP_016874096.1
13
1199
XM_017018608.1
XP_016874097.1
15
1569
XM_017018609.1
XP_016874098.1
15
1860
XM_017018610.1
XP_016874099.1
14
1744
XM_017018611.1
XP_016874100.1
15
1795
XM_017018612.1
XP_016874101.1
15
1982
XM_017018613.1
XP_016874102.1
14
1776
XM_017018614.1
XP_016874103.1
15
1767
XM_017018615.1
XP_016874104.1
14
1866
XM_017018616.1
XP_016874105.1
13
1660
XM_017018617.1
XP_016874106.1
15
1865
XM_017018618.1
XP_016874107.1
17
2181
XM_017018619.1
XP_016874108.1
15
1966
XM_017018620.1
XP_016874109.1
15
2026
XM_017018621.1
XP_016874110.1
14
1910
XM_017018622.1
XP_016874111.1
14
1867
GenBank mRNA
AB014552.1
-
14
1480
AK023867.1
-
14
1478
AK223603.1
-
15
1575
AK289641.1
-
15
1403
AK293109.1
-
14
1444
AK294110.1
-
12
1320
AK296526.1
-
7
687
AK315925.1
-
7
614
BC001331.1
-
14
1174
3458359
BC002378.2
-
14
1444
2961127
BC006191.2
-
14
1486
3936851
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0