Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751518.3 | XP_003751566.1 | 10 | 1569 | ||
XM_006252367.2 | XP_006252429.1 | 10 | 1607 | |||
XM_006252368.2 | XP_006252430.1 | 9 | 1459 | |||
XM_008770927.1 | XP_008769149.1 | 10 | 1298 |