Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044290.2 | NP_001037755.2 | 4 | 421 | ||
GenBank mRNA | BC083898.1 | - | 4 | 358 | 7133376 | |
FM031800.1 | - | 4 | 421 |